Assay Details
Target Gene Details
Entrez Gene ID: | 7170 |
Gene Name: | tropomyosin 3 |
Gene Aliases: |
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30 |
Location: |
Chr.1:154155304-154192135 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520398 | Chr.1:154095720 - 154169432 on Build GRCh38 | Gain+Loss | NUP210L TPM3 MIR5698 |
nsv1000153 | Chr.1:154101773 - 154179941 on Build GRCh38 | Gain | NUP210L TPM3 MIR5698 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |