Assay Details
Target Gene Details
Entrez Gene ID: | 8743 |
Gene Name: | tumor necrosis factor superfamily member 10 |
Gene Aliases: |
APO2L, Apo-2L, CD253, TL2, TNLG6A, TRAIL |
Location: |
Chr.3:172505508-172523507 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFSF10 | NM_001190942.1 | NP_001177871.1 | ||
NM_003810.3 | NP_003801.1 | |||
NR_033994.1 | ||||
AK296085.1 | ||||
AK312742.1 | ||||
AY312579.1 | AAR16184.1 | |||
BC032722.1 | AAH32722.1 | |||
BT019564.1 | AAV38371.1 | |||
CR456895.1 | CAG33176.1 | |||
DA041872.1 | ||||
DA969653.1 | ||||
DQ848564.1 | ||||
EU183231.1 | ||||
U37518.1 | AAC50332.1 | |||
U57059.1 | AAB01233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829794 | Chr.3:172441561 - 172607903 on Build GRCh38 | Gain | GHSR TNFSF10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |