Assay Details
Target Gene Details
Entrez Gene ID: | 11201 |
Gene Name: | DNA polymerase iota |
Gene Aliases: |
RAD30B, RAD3OB |
Location: |
Chr.18:54269479-54321266 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLI | NM_007195.2 | NP_009126.2 | ||
XM_005258188.3 | XP_005258245.1 | |||
XM_005258192.4 | XP_005258249.1 | |||
XM_006722378.2 | XP_006722441.1 | |||
XM_011525796.2 | XP_011524098.1 | |||
XM_011525797.1 | XP_011524099.1 | |||
AF140501.1 | AAD50381.1 | |||
AF245438.1 | AAF63383.1 | |||
AK093688.1 | 1 | 1800 | ||
AK301578.1 | ||||
AL136670.1 | CAB66605.1 | |||
BC032662.1 | AAH32662.1 | |||
DB450447.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |