Assay Details
Target Gene Details
Entrez Gene ID: | 60437 |
Gene Name: | cadherin 26 |
Gene Aliases: |
VR20 |
Location: |
Chr.20:59957764-60035629 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH26 | NM_177980.2 | NP_817089.1 | ||
XM_011528969.2 | XP_011527271.1 | |||
XM_017027994.1 | 1 | 667 | XP_016883483.1 | |
XM_017027995.1 | 1 | 668 | XP_016883484.1 | |
XM_017027996.1 | 1 | 668 | XP_016883485.1 | |
XM_017027997.1 | 1 | 668 | XP_016883486.1 | |
XM_017027998.1 | 1 | 665 | XP_016883487.1 | |
XM_017027999.1 | 1 | 665 | XP_016883488.1 | |
XM_017028000.1 | XP_016883489.1 | |||
XM_017028001.1 | 1 | 665 | XP_016883490.1 | |
XM_017028002.1 | 1 | 666 | XP_016883491.1 | |
AK055202.1 | ||||
DB218314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1009769 | Chr.20:59957379 - 59963538 on Build GRCh38 | Deletion | CDH26 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |