Assay Details
Target Gene Details
Entrez Gene ID: | 4711 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B5 |
Gene Aliases: |
CISGDH, SGDH |
Location: |
Chr.3:179604787-179624500 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFB5 | NM_001199957.1 | NP_001186886.1 | ||
NM_001199958.1 | NP_001186887.1 | |||
NM_002492.3 | NP_002483.1 | |||
AF047181.1 | AAC04266.1 | |||
AK313822.1 | ||||
AW955118.1 | ||||
BC005271.1 | AAH05271.1 | |||
BC009796.1 | AAH09796.1 | |||
BC017909.1 | ||||
BC093071.1 | AAH93071.1 | |||
BG776403.1 | ||||
BX648034.1 | ||||
DA112751.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |