Assay Details
Target Gene Details
Entrez Gene ID: | 3359 |
Gene Name: | 5-hydroxytryptamine receptor 3A |
Gene Aliases: |
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3 |
Location: |
Chr.11:113975075-113990313 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR3A | NM_000869.5 | NP_000860.2 | ||
NM_001161772.2 | NP_001155244.1 | |||
NM_213621.3 | 7 | 1287 | NP_998786.2 | |
NR_046363.1 | ||||
AF498984.1 | AAM21131.1 | |||
AJ003078.1 | 7 | 1076 | CAA05851.1 | |
AJ003079.1 | CAA05852.1 | |||
AJ003080.1 | CAA05853.1 | |||
AK299973.1 | ||||
AK304630.1 | ||||
AK313896.1 | ||||
BC002354.2 | AAH02354.1 | |||
BC004453.1 | AAH04453.2 | |||
BT007204.1 | AAP35868.1 | |||
CD014118.1 | 7 | 1174 | ||
D49394.1 | BAA08387.1 | |||
S82612.1 | AAB37533.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3627783 | Chr.11:113969123 - 113990232 on Build GRCh38 | Loss | HTR3A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |