Assay Details
Target Gene Details
Entrez Gene ID: | 8829 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
BDCA4, CD304, NP1, NRP, VEGF165R |
Location: |
Chr.10:33177491-33334905 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRP1 | NM_001244972.1 | 17 | 4205 | NP_001231901.1 |
NM_001244973.1 | 17 | 4202 | NP_001231902.1 | |
NM_003873.5 | 17 | 4223 | NP_003864.4 | |
NR_045259.1 | 16 | 4020 | ||
XM_006717521.1 | 17 | 4000 | XP_006717584.1 | |
XM_006717522.1 | 16 | 3949 | XP_006717585.1 | |
XM_006717523.2 | 16 | 3946 | XP_006717586.1 | |
XM_006717524.1 | 16 | 3895 | XP_006717587.1 | |
XM_006717525.1 | 16 | 3892 | XP_006717588.1 | |
XM_017016865.1 | 16 | 3925 | XP_016872354.1 | |
XM_017016866.1 | 16 | 3817 | XP_016872355.1 | |
AB209641.1 | 17 | 3851 | BAD92878.1 | |
AL833608.1 | 16 | 3751 | CAH10397.1 | |
BX510902.1 | 17 | 3887 | CAD91133.1 | |
BX648250.1 | 17 | 4171 | ||
CR749333.1 | 17 | 4224 | CAH18187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss | NRP1 LOC105376488 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |