Assay Details
Target Gene Details
Entrez Gene ID: | 216 |
Gene Name: | aldehyde dehydrogenase 1 family member A1 |
Gene Aliases: |
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1 |
Location: |
Chr.9:72900662-72953317 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH1A1 | NM_000689.4 | 10 | 1472 | NP_000680.2 |
AB209821.1 | 10 | 1199 | BAD93058.1 | |
AF003341.1 | 10 | 1155 | AAC51652.1 | |
AK000118.1 | 10 | 1199 | ||
AK026641.1 | 10 | 1199 | ||
AK293176.1 | 10 | 1165 | ||
AK302168.1 | 9 | 983 | ||
AY390731.1 | 10 | 1155 | AAR92229.1 | |
BC001505.2 | 10 | 1180 | AAH01505.1 | |
BT006921.1 | 10 | 1155 | AAP35567.1 | |
EU668318.1 | 10 | 1199 | ||
EU794589.1 | 10 | 1155 | ||
FJ224286.1 | 10 | 1208 | ||
K03000.1 | 6 | 672 | AAA51695.1 | |
M26761.1 | 6 | 672 | AAA35518.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |