Assay Details
Target Gene Details
Entrez Gene ID: | 119504 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
APC16, C10orf104, CENP-27, MSAG, bA570G20.3 |
Location: |
Chr.10:72216000-72235860 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANAPC16 | NM_001242546.1 | 5 | 763 | NP_001229475.1 |
NM_001242547.1 | 4 | 371 | NP_001229476.1 | |
NM_001242548.1 | 3 | 296 | NP_001229477.1 | |
NM_173473.3 | 4 | 437 | NP_775744.1 | |
NR_038391.1 | 3 | 202 | ||
NR_038392.1 | 3 | 268 | ||
BC009530.1 | 4 | 389 | AAH09530.1 | |
BM768948.1 | 3 | 240 | ||
CB984695.1 | 3 | 276 | ||
CD511334.1 | 4 | 364 | ||
DA310509.1 | 3 | 202 | ||
DA739927.1 | 4 | 437 | ||
EU552924.1 | 4 | 410 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |