Assay Details
Target Gene Details
Entrez Gene ID: | 10099 |
Gene Name: | tetraspanin 3 |
Gene Aliases: |
TM4-A, TM4SF8, TSPAN-3 |
Location: |
Chr.15:77044017-77071228 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN3 | NM_001168412.1 | 6 | 1007 | NP_001161884.1 |
NM_005724.5 | 7 | 1199 | NP_005715.1 | |
NM_198902.2 | 6 | 1124 | NP_944492.1 | |
XM_017021857.1 | 7 | 808 | XP_016877346.1 | |
AF054840.1 | AAC69716.1 | |||
AF133423.1 | 7 | 999 | AAF08362.1 | |
AK001305.1 | 7 | 1045 | BAA91613.1 | |
AK001310.1 | 7 | 1031 | BAA91615.1 | |
AK001326.1 | 7 | 1142 | BAA91627.1 | |
AK027793.1 | 4 | 2250 | ||
AK075336.1 | 7 | 1080 | ||
AK075482.1 | 6 | 1005 | ||
AK298157.1 | 6 | 888 | ||
BC000704.2 | 7 | 1100 | AAH00704.1 | |
BC004280.1 | 7 | 1061 | AAH04280.1 | |
BC009248.2 | 7 | 1064 | AAH09248.1 | |
BC011206.1 | 7 | 1093 | AAH11206.1 | |
M69023.1 | 3 | 473 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517068 | Chr.15:76962203 - 77088880 on Build GRCh38 | Gain+Loss | TSPAN3 PSTPIP1 |
esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain | SCAPER TSPAN3 RCN2 PSTPIP1 |
nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss | TSPAN3 RCN2 PEAK1 PSTPIP1 |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion | NRG4 TSPAN3 MIR3713 LOC645752 RCN2 FBXO22-AS1 UBE2Q2 PEAK1 LOC101929439 PSTPIP1 ETFA TMEM266 ISL2 LINGO1-AS1 SCAPER FBXO22 TYRO3P LINGO1 LINGO1-AS2 LOC101929457 HMG20A LOC441728 LINC00597 |
nsv1048833 | Chr.15:77002252 - 77046918 on Build GRCh38 | Gain | TSPAN3 PSTPIP1 |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain | LINGO1-AS1 SCAPER TSPAN3 LINGO1 RCN2 LINGO1-AS2 PEAK1 LOC101929457 HMG20A LINC00597 PSTPIP1 |
nsv1622 | Chr.15:77022478 - 77051924 on Build GRCh38 | Insertion | TSPAN3 PSTPIP1 |
nsv509576 | Chr.15:76962019 - 77047336 on Build GRCh38 | Insertion | TSPAN3 PSTPIP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |