Assay Details
Target Gene Details
Entrez Gene ID: | 101929664 |
Gene Name: | HORMAD2 antisense RNA 1 |
Gene Aliases: |
MTMR3-AS1 |
Location: |
Chr.22:30008742-30080480 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HORMAD2-AS1 | NR_110541.1 | |||
BG618381.1 |
Target Gene Details
Entrez Gene ID: | 8897 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
FYVE-DSP1, ZFYVE10 |
Location: |
Chr.22:29883165-30030868 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTMR3 | NM_021090.3 | 20 | 4499 | NP_066576.1 |
NM_153050.2 | 20 | 4415 | NP_694690.1 | |
NM_153051.2 | 19 | 4388 | NP_694691.1 | |
XM_005261803.2 | 22 | 4644 | XP_005261860.1 | |
XM_005261804.3 | 21 | 4561 | XP_005261861.1 | |
XM_005261805.2 | 21 | 4617 | XP_005261862.1 | |
XM_005261806.2 | 21 | 4526 | XP_005261863.1 | |
XM_005261808.2 | 20 | 4506 | XP_005261865.1 | |
XM_017029028.1 | 20 | 4418 | XP_016884517.1 | |
XM_017029029.1 | 19 | 4381 | XP_016884518.1 | |
XM_017029030.1 | 19 | 4313 | XP_016884519.1 | |
XM_017029031.1 | 18 | 4274 | XP_016884520.1 | |
XM_017029032.1 | 19 | 4271 | XP_016884521.1 | |
XM_017029033.1 | 15 | 3845 | XP_016884522.1 | |
XM_017029034.1 | 21 | 5848 | XP_016884523.1 | |
AB002369.1 | 20 | 4418 | ||
BC148216.1 | 18 | 4207 | ||
BC151217.1 | 20 | 4513 | ||
BC152455.1 | 20 | 4418 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |