Assay Details
Target Gene Details
Entrez Gene ID: | 22850 |
Gene Name: | ADNP homeobox 2 |
Gene Aliases: |
ZNF508 |
Location: |
Chr.18:80109031-80140346 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADNP2 | NM_014913.3 | 4 | 1698 | NP_055728.1 |
XM_005266656.4 | 4 | 1552 | XP_005266713.1 | |
XM_011525883.2 | 4 | 1412 | XP_011524185.1 | |
AB020670.1 | 4 | 1428 | ||
AK022688.1 | 1 | 217 | BAB14180.1 | |
AK292566.1 | 4 | 1467 | ||
BC071589.1 | 4 | 1698 | AAH71589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3413n100 | Chr.18:80023442 - 80257174 on Build GRCh38 | Gain | RBFA TXNL4A RBFADN PARD6G-AS1 PARD6G ADNP2 |
nsv428359 | Chr.18:79277548 - 80255221 on Build GRCh38 | Gain | RBFA TXNL4A LOC284240 ATP9B NFATC1 LOC107985149 LOC284241 CTDP1 KCNG2 ADNP2 PQLC1 RBFADN HSBP1L1 PARD6G-AS1 PARD6G LOC105376875 |
nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain | RBFA TXNL4A LOC284240 ATP9B NFATC1 LOC107985149 LOC284241 CTDP1 LOC100996274 KCNG2 ADNP2 PQLC1 SALL3 RBFADN HSBP1L1 PARD6G-AS1 PARD6G LOC105376875 |
esv3643330 | Chr.18:80099479 - 80182196 on Build GRCh38 | Loss | PARD6G-AS1 PARD6G ADNP2 |
dgv3414n100 | Chr.18:80110621 - 80257174 on Build GRCh38 | Gain | PARD6G-AS1 PARD6G ADNP2 |
esv3643332 | Chr.18:80136109 - 80224214 on Build GRCh38 | Loss | PARD6G-AS1 PARD6G ADNP2 |
nsv1160552 | Chr.18:80135813 - 80222390 on Build GRCh38 | Duplication | PARD6G-AS1 PARD6G ADNP2 |
esv3643326 | Chr.18:79799861 - 80157032 on Build GRCh38 | Loss | RBFA TXNL4A RBFADN HSBP1L1 PARD6G-AS1 KCNG2 ADNP2 PQLC1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |