Assay Details
Target Gene Details
Entrez Gene ID: | 10718 |
Gene Name: | neuregulin 3 |
Gene Aliases: |
HRG3, pro-NRG3 |
Location: |
Chr.10:81875298-82989979 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRG3 | NM_001010848.3 | 1 | 226 | NP_001010848.2 |
NM_001165972.1 | 1 | 226 | NP_001159444.1 | |
XM_005269444.4 | 1 | 227 | XP_005269501.1 | |
XM_011539172.2 | 1 | 229 | XP_011537474.1 | |
XM_011539173.2 | 1 | 229 | XP_011537475.1 | |
XM_011539175.2 | 1 | 229 | XP_011537477.1 | |
XM_017015573.1 | 1 | 231 | XP_016871062.1 | |
XM_017015574.1 | 1 | 229 | XP_016871063.1 | |
XM_017015575.1 | 1 | 231 | XP_016871064.1 | |
XM_017015576.1 | 1 | 229 | XP_016871065.1 | |
XM_017015577.1 | 1 | 231 | XP_016871066.1 | |
XM_017015578.1 | 1 | 229 | XP_016871067.1 | |
XM_017015579.1 | 1 | 230 | XP_016871068.1 | |
XM_017015580.1 | 1 | 229 | XP_016871069.1 | |
XM_017015581.1 | 1 | 229 | XP_016871070.1 | |
XM_017015584.1 | 1 | 242 | XP_016871073.1 | |
AK296563.1 | 1 | 48 | ||
BC136811.1 | 1 | 226 | ||
DQ001411.1 | 1 | 199 | AAY17216.1 | |
HM068873.1 | 1 | 199 | ||
HM068874.1 | 1 | 199 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551669 | Chr.10:81374678 - 82080786 on Build GRCh38 | Gain | NRG3 |
esv3891873 | Chr.10:81025608 - 82340753 on Build GRCh38 | Gain | NRG3 NRG3-AS1 |
nsv831925 | Chr.10:81771475 - 81938851 on Build GRCh38 | Gain | NRG3 |
nsv551670 | Chr.10:81545860 - 81972366 on Build GRCh38 | Loss | NRG3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |