Assay Details
Target Gene Details
Entrez Gene ID: | 79368 |
Gene Name: | Fc receptor like 2 |
Gene Aliases: |
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C |
Location: |
Chr.1:157745733-157777472 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL2 | NM_001159488.1 | 9 | 1522 | NP_001152960.1 |
NM_030764.3 | 12 | 1703 | NP_110391.2 | |
NR_125358.1 | 8 | 785 | ||
XM_011509974.2 | 12 | 2064 | XP_011508276.1 | |
XM_011509975.2 | 11 | 2000 | XP_011508277.1 | |
XM_011509976.2 | 11 | 1973 | XP_011508278.1 | |
XM_017002316.1 | 11 | 2009 | XP_016857805.1 | |
XM_017002317.1 | 11 | 1812 | XP_016857806.1 | |
XM_017002318.1 | 10 | 1732 | XP_016857807.1 | |
XM_017002319.1 | 10 | 1724 | XP_016857808.1 | |
AF319438.1 | 7 | 1188 | AAK01402.1 | |
AF459633.1 | 12 | 1691 | AAL60249.1 | |
AL833194.1 | 7 | 3562 | ||
AL833361.1 | 2 | 2223 | ||
AY043465.1 | 12 | 1690 | AAK91778.1 | |
AY358130.1 | 12 | 1693 | AAQ88497.1 | |
BC069185.1 | 8 | 789 | AAH69185.1 | |
BC129836.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |