Assay Details
Target Gene Details
Entrez Gene ID: | 10397 |
Gene Name: | N-myc downstream regulated 1 |
Gene Aliases: |
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5 |
Location: |
Chr.8:133237171-133297587 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDRG1 | NM_001135242.1 | 16 | 2659 | NP_001128714.1 |
NM_001258432.1 | 14 | 2145 | NP_001245361.1 | |
NM_001258433.1 | 15 | 2156 | NP_001245362.1 | |
NM_006096.3 | 16 | 2262 | NP_006087.2 | |
XM_011516791.1 | 16 | 2596 | XP_011515093.1 | |
XM_011516792.1 | 9 | 2495 | XP_011515094.1 | |
AF004162.1 | 15 | 2101 | AAC13419.1 | |
AK091147.1 | 15 | 2105 | ||
AK091269.1 | ||||
AK095649.1 | 14 | 1998 | BAC04597.1 | |
AK096562.1 | 16 | 2097 | ||
AK124709.1 | 8 | 3169 | ||
AK126924.1 | 16 | 2941 | ||
AK129867.1 | 7 | 1536 | ||
BC003175.1 | 16 | 2190 | AAH03175.1 | |
D87953.1 | 16 | 2195 | BAA13505.1 | |
X92845.1 | 16 | 2183 | CAA63430.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79281324] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |