Assay Details
Target Gene Details
Entrez Gene ID: | 1240 |
Gene Name: | chemerin chemokine-like receptor 1 |
Gene Aliases: |
CHEMERINR, ChemR23, DEZ, RVER1 |
Location: |
Chr.12:108288044-108339348 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMKLR1 | NM_001142343.1 | 4 | 689 | NP_001135815.1 |
NM_001142344.1 | 3 | 476 | NP_001135816.1 | |
NM_001142345.1 | 3 | 303 | NP_001135817.1 | |
NM_004072.2 | 3 | 613 | NP_004063.1 | |
XM_017018820.1 | 2 | 431 | XP_016874309.1 | |
AB112777.1 | 1 | 106 | ||
AK291800.1 | 3 | 470 | ||
AY497547.1 | 1 | 106 | AAR90850.1 | |
BC106927.2 | 4 | 503 | AAI06928.1 | |
BC106928.1 | 4 | 503 | AAI06929.1 | |
BP328503.1 | 3 | 303 | ||
BT019556.1 | 1 | 106 | AAV38363.1 | |
BT019557.1 | 1 | 106 | AAV38364.1 | |
DA466082.1 | 3 | 476 | ||
DA939526.1 | 3 | 284 | ||
U79526.1 | 3 | 466 | AAC51258.1 | |
U79527.1 | 3 | 619 | AAC51259.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |