Assay Details
Target Gene Details
Entrez Gene ID: | 9570 |
Gene Name: | golgi SNAP receptor complex member 2 |
Gene Aliases: |
Bos1, EPM6, GS27 |
Location: |
Chr.17:46923086-46975519 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2656635 | Chr.17:46204086 - 47091135 on Build GRCh38 | Deletion | NSFP1 KANSL1 MIR5089 GOSR2 LOC107987253 LRRC37A WNT9B LRRC37A2 RPRML ARL17B ARL17A NSF WNT3 LOC107987254 |
esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss | NSFP1 KANSL1 MIR5089 GOSR2 KANSL1-AS1 LOC107987253 LRRC37A WNT9B RPS2P47 MAPT LRRC37A2 RPRML ARL17B ARL17A NSF WNT3 LOC107987254 CDC27 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |