Assay Details
Target Gene Details
Entrez Gene ID: | 285755 |
Gene Name: | peptidylprolyl isomerase like 6 |
Gene Aliases: |
PPIase, RSPH12, bA425D10.6, dJ919F19.1 |
Location: |
Chr.6:109390215-109441171 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIL6 | NM_001111298.2 | 9 | 1534 | NP_001104768.2 |
NM_001286360.1 | 7 | 1360 | NP_001273289.1 | |
NM_001286361.1 | 6 | 1224 | NP_001273290.1 | |
NM_173672.4 | 8 | 1456 | NP_775943.1 | |
NR_104429.1 | 9 | 1576 | ||
XM_011535765.2 | 8 | 898 | XP_011534067.1 | |
XM_017010774.1 | 7 | 788 | XP_016866263.1 | |
AK293674.1 | 9 | 1028 | ||
AK315790.1 | 8 | 929 | ||
BC038716.1 | 8 | 915 | AAH38716.1 | |
BI824055.1 | ||||
DQ363562.1 | 8 | 1456 | ||
DQ423529.1 | 9 | 1534 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |