Assay Details
Target Gene Details
Entrez Gene ID: | 220002 |
Gene Name: | cytochrome b561 family member A3 |
Gene Aliases: |
CYBASC3, LCYTB |
Location: |
Chr.11:61348748-61362299 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYB561A3 | NM_001161454.1 | 1 | 364 | NP_001154926.1 |
NM_001300763.1 | 1 | 364 | NP_001287692.1 | |
NM_153611.4 | 1 | 364 | NP_705839.3 | |
XM_011544821.1 | 1 | 380 | XP_011543123.1 | |
XM_011544823.1 | 1 | 380 | XP_011543125.1 | |
XM_011544824.1 | 1 | 380 | XP_011543126.1 | |
XM_017017345.1 | 1 | 380 | XP_016872834.1 | |
XM_017017346.1 | 1 | 380 | XP_016872835.1 | |
AK056751.1 | 1 | 355 | ||
BC004391.1 | 1 | 338 | AAH04391.1 | |
BC014045.2 | 1 | 174 | AAH14045.2 | |
CF125719.1 | 1 | 364 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |