Assay Details
Target Gene Details
Entrez Gene ID: | 387338 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 4 |
Gene Aliases: |
SHTAP |
Location: |
Chr.1:46340177-46365152 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN4 | NM_001256127.1 | 4 | 1280 | NP_001243056.1 |
NM_001256128.1 | 4 | 912 | NP_001243057.1 | |
NM_199044.3 | 4 | 1335 | NP_950245.2 | |
NR_045789.1 | 5 | 1510 | ||
NR_045790.1 | 3 | 1180 | ||
NR_045791.1 | 3 | 998 | ||
AK021577.1 | BAB13847.1 | |||
AK057420.1 | 4 | 1271 | ||
AK097524.1 | 4 | 1312 | ||
AK128066.1 | 4 | 3051 | BAC87258.1 | |
AK291741.1 | 4 | 708 | ||
AK292353.1 | 4 | 886 | ||
AK295003.1 | 5 | 880 | ||
BC014441.1 | 4 | 756 | AAH14441.1 | |
BX360729.2 | 4 | 767 | ||
BX648592.1 | 3 | 569 | ||
DA799314.1 | 3 | 372 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577837 | Chr.1:46338361 - 46357126 on Build GRCh38 | Loss |
![]() |
nsv159955 | Chr.1:46347871 - 46354655 on Build GRCh38 | Deletion |
![]() |
nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)