Assay Details
Target Gene Details
Entrez Gene ID: | 4688 |
Gene Name: | neutrophil cytosolic factor 2 |
Gene Aliases: |
NCF-2, NOXA2, P67-PHOX, P67PHOX |
Location: |
Chr.1:183555562-183590921 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCF2 | NM_000433.3 | 15 | 2036 | NP_000424.2 |
NM_001127651.2 | 16 | 1873 | NP_001121123.1 | |
NM_001190789.1 | 13 | 1793 | NP_001177718.1 | |
NM_001190794.1 | 14 | 1901 | NP_001177723.1 | |
XM_005245207.1 | 14 | 1857 | XP_005245264.1 | |
XM_011509580.1 | 16 | 1874 | XP_011507882.1 | |
XM_011509581.1 | 16 | 1861 | XP_011507883.1 | |
AB209647.1 | 13 | 5306 | BAD92884.1 | |
AF527950.1 | 15 | 1783 | AAM89263.1 | |
AK296672.1 | ||||
AW951155.1 | 1 | 199 | ||
BC001606.1 | 15 | 2013 | AAH01606.1 | |
BM979513.1 | 1 | 394 | ||
BQ029626.1 | 1 | 393 | ||
BU617086.1 | 1 | 394 | ||
M32011.1 | 16 | 1828 | AAA36379.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |