Assay Details
Target Gene Details
Entrez Gene ID: | 26054 |
Gene Name: | SUMO1/sentrin specific peptidase 6 |
Gene Aliases: |
SSP1, SUSP1 |
Location: |
Chr.6:75601509-75718285 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SENP6 | NM_001100409.2 | 7 | 1584 | NP_001093879.1 |
NM_001304792.1 | 7 | 1584 | NP_001291721.1 | |
NM_015571.3 | 8 | 1605 | NP_056386.2 | |
XM_006715457.3 | 9 | 1359 | XP_006715520.1 | |
XM_011535746.2 | 7 | 1157 | XP_011534048.1 | |
XM_011535747.2 | 7 | 1508 | XP_011534049.1 | |
XM_017010719.1 | 8 | 1247 | XP_016866208.1 | |
XM_017010720.1 | 8 | 563 | XP_016866209.1 | |
XM_017010721.1 | 8 | 571 | XP_016866210.1 | |
XM_017010722.1 | 9 | 1378 | XP_016866211.1 | |
XM_017010723.1 | 9 | 1355 | XP_016866212.1 | |
XM_017010724.1 | 9 | 1730 | XP_016866213.1 | |
XM_017010725.1 | 6 | 1490 | XP_016866214.1 | |
AB018340.2 | 8 | 633 | ||
AF196304.1 | 8 | 589 | AAF04852.1 | |
AF306508.1 | 8 | 604 | AAG30253.1 | |
AF307849.1 | 8 | 664 | AAG29831.1 | |
AK001406.1 | 5 | 259 | ||
AK096455.1 | 7 | 1166 | BAC04794.1 | |
AK126921.1 | 7 | 1584 | ||
BC028583.1 | 7 | 791 | AAH28583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1032961 | Chr.6:75633859 - 76079135 on Build GRCh38 | Gain | MYO6 SENP6 IMPG1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |