Assay Details
Target Gene Details
Entrez Gene ID: | 5832 |
Gene Name: | aldehyde dehydrogenase 18 family member A1 |
Gene Aliases: |
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B |
Location: |
Chr.10:95605929-95656810 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH18A1 | NM_001017423.1 | 13 | 1754 | NP_001017423.1 |
NM_001323412.1 | 12 | 1545 | NP_001310341.1 | |
NM_001323413.1 | 13 | 1911 | NP_001310342.1 | |
NM_001323414.1 | 13 | 1780 | NP_001310343.1 | |
NM_001323415.1 | 13 | 1905 | NP_001310344.1 | |
NM_001323416.1 | 12 | 1696 | NP_001310345.1 | |
NM_001323417.1 | 12 | 1655 | NP_001310346.1 | |
NM_001323418.1 | 12 | 1690 | NP_001310347.1 | |
NM_001323419.1 | 10 | 1290 | NP_001310348.1 | |
NM_002860.3 | 13 | 1760 | NP_002851.2 | |
AK225455.1 | 12 | 1330 | ||
AK225465.1 | 12 | 1330 | ||
AK226032.1 | 12 | 1330 | ||
AK295487.1 | 12 | 1411 | ||
AK299557.1 | 10 | 1188 | ||
AK312271.1 | 13 | 1662 | ||
BC106054.1 | 13 | 1595 | AAI06055.1 | |
BC117240.1 | 13 | 1701 | ||
BC117242.1 | 13 | 1701 | ||
BC143930.1 | 13 | 1695 | ||
U68758.1 | 13 | 1569 | AAD00169.1 | |
U76542.1 | 13 | 1575 | AAD17454.1 | |
X94453.1 | 13 | 1657 | CAA64224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526180 | Chr.10:95572438 - 95751280 on Build GRCh38 | Gain | ALDH18A1 ENTPD1 TCTN3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |