Assay Details
Target Gene Details
Entrez Gene ID: | 1609 |
Gene Name: | diacylglycerol kinase theta |
Gene Aliases: |
DAGK, DAGK4, DAGK7 |
Location: |
Chr.4:958884-973560 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DGKQ | NM_001347.3 | NP_001338.2 | ||
XM_011513411.1 | XP_011511713.1 | |||
XM_011513412.1 | XP_011511714.1 | |||
XM_011513414.2 | XP_011511716.1 | |||
XM_011513415.1 | XP_011511717.1 | |||
XM_017007814.1 | XP_016863303.1 | |||
XM_017007815.1 | XP_016863304.1 | |||
AB209503.1 | BAD92740.1 | |||
BC063801.1 | AAH63801.1 | |||
GU727647.1 | ||||
L38707.1 | AAA98749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain | CPLX1 TMEM175 FGFRL1 IDUA GAK SLC26A1 RNF212 DGKQ LOC100129917 |
nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss | TMEM175 FGFRL1 IDUA SLC26A1 DGKQ |
nsv4196 | Chr.4:961755 - 1004599 on Build GRCh38 | Deletion | IDUA SLC26A1 DGKQ |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | LOC101928548 ATP5I UVSSA LOC100129917 TMEM175 CTBP1-AS LOC100130872 SPON2 CPLX1 CRIPAK PCGF3 PDE6B IDUA TMED11P RNF212 LOC107986211 LOC101928521 MFSD7 FGFRL1 CTBP1-AS2 NKX1-1 GAK SLC26A1 MYL5 CTBP1 LOC105374344 MAEA DGKQ |
nsv950327 | Chr.4:945913 - 978512 on Build GRCh38 | Deletion | TMEM175 DGKQ |
dgv5079n100 | Chr.4:843485 - 970603 on Build GRCh38 | Gain | TMEM175 GAK DGKQ |
esv29984 | Chr.4:857569 - 970571 on Build GRCh38 | Loss | TMEM175 GAK DGKQ |
nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss | CPLX1 TMEM175 FGFRL1 IDUA GAK SLC26A1 RNF212 DGKQ |
nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain | CPLX1 TMEM175 FGFRL1 IDUA GAK SLC26A1 DGKQ |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |