Assay Details
Target Gene Details
Entrez Gene ID: | 80067 |
Gene Name: | DDB1 and CUL4 associated factor 17 |
Gene Aliases: |
C2orf37 |
Location: |
Chr.2:171434166-171491029 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF17 | NM_001164821.1 | 1 | 99 | NP_001158293.1 |
NM_025000.3 | 1 | 99 | NP_079276.2 | |
NR_028482.1 | 1 | 99 | ||
XM_006712767.1 | XP_006712830.1 | |||
XM_011511881.1 | 1 | 94 | XP_011510183.1 | |
XM_011511882.1 | 1 | 94 | XP_011510184.1 | |
XM_017004995.1 | 1 | 94 | XP_016860484.1 | |
XM_017004996.1 | 1 | 94 | XP_016860485.1 | |
XM_017004997.1 | 1 | 94 | XP_016860486.1 | |
XM_017004998.1 | XP_016860487.1 | |||
XM_017004999.1 | 1 | 94 | XP_016860488.1 | |
XM_017005000.1 | 1 | 94 | XP_016860489.1 | |
XM_017005001.1 | 1 | 94 | XP_016860490.1 | |
XM_017005002.1 | XP_016860491.1 | |||
AK023158.1 | BAB14436.1 | |||
AK307877.1 | 1 | 99 | ||
CR933646.1 | CAI45947.1 |
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
METTL8 | NM_001321157.1 | NP_001308086.1 | ||
NM_001321158.1 | NP_001308087.1 | |||
NM_001321161.1 | NP_001308090.1 | |||
NM_024770.4 | NP_079046.2 | |||
NR_135568.1 | ||||
XM_011511875.2 | XP_011510177.1 | |||
XM_017004984.1 | XP_016860473.1 | |||
XM_017004986.1 | XP_016860475.1 | |||
XM_017004987.1 | XP_016860476.1 | |||
AK292108.1 | ||||
AK297141.1 | ||||
DB031573.1 | ||||
DC349499.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |