Assay Details
Target Gene Details
Entrez Gene ID: | 8924 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase 2 |
Gene Aliases: |
D15F37S1, MRT38, SHEP1, jdf2, p528 |
Location: |
Chr.15:28111037-28322173 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 87 - Exon 87 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HERC2 | NM_004667.5 | NP_004658.3 | ||
XM_005268276.4 | XP_005268333.1 | |||
XM_006720726.3 | XP_006720789.1 | |||
XM_006720727.3 | XP_006720790.1 | |||
XM_011522131.2 | XP_011520433.1 | |||
XM_011522133.2 | XP_011520435.1 | |||
XM_017022695.1 | XP_016878184.1 | |||
XM_017022696.1 | XP_016878185.1 | |||
XM_017022697.1 | XP_016878186.1 | |||
XM_017022698.1 | XP_016878187.1 | |||
AF071172.3 | AAD08657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635988 | Chr.15:28098445 - 28194723 on Build GRCh38 | Gain | HERC2 OCA2 |
nsv428299 | Chr.15:28042424 - 28817373 on Build GRCh38 | Loss | HERC2 LOC100289656 LOC645202 PDCD6IPP2 WHAMMP2 LOC100132202 MIR4509-2 GOLGA8G OCA2 LOC101929047 HERC2P11 MIR4509-3 GOLGA8F GOLGA8M HERC2P9 |
nsv568663 | Chr.15:27783102 - 28176880 on Build GRCh38 | Loss | HERC2 OCA2 |
nsv1039684 | Chr.15:28080438 - 28315658 on Build GRCh38 | Gain | HERC2 OCA2 |
nsv1052131 | Chr.15:27932591 - 28460005 on Build GRCh38 | Gain | HERC2 MIR4509-2 LOC645202 OCA2 GOLGA8F |
nsv1038472 | Chr.15:27647423 - 28256473 on Build GRCh38 | Gain | HERC2 OCA2 |
nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain | HERC2 LOC100289656 LOC645202 PDCD6IPP2 WHAMMP2 LOC107984752 LOC100132202 MIR4509-2 GOLGA8G GOLGA6L7P OCA2 APBA2 LOC101929047 HERC2P11 MIR4509-3 GOLGA8F GOLGA8M HERC2P9 |
nsv1047941 | Chr.15:28110984 - 28168961 on Build GRCh38 | Gain | HERC2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |