Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | 12 | 1604 | NP_001137498.1 |
NM_001144027.2 | 11 | 1502 | NP_001137499.1 | |
NM_001144028.2 | 12 | 1583 | NP_001137500.1 | |
NM_014434.3 | 12 | 1604 | NP_055249.1 | |
XM_011518545.1 | 11 | 1492 | XP_011516847.1 | |
XM_011518547.1 | 12 | 1394 | XP_011516849.1 | |
XM_011518548.2 | 11 | 2794 | XP_011516850.1 | |
XM_017014626.1 | 12 | 1604 | XP_016870115.1 | |
XM_017014627.1 | 12 | 1604 | XP_016870116.1 | |
AF199509.1 | 12 | 1540 | AAF25205.1 | |
AK026089.1 | 12 | 1576 | ||
AK074403.1 | 11 | 1463 | ||
AK225995.1 | 12 | 1575 | ||
AK290026.1 | 12 | 1601 | ||
AY077845.1 | 12 | 1522 | AAL77754.1 | |
BC015735.1 | 12 | 1540 | AAH15735.1 | |
BC093782.1 | 12 | 1521 | AAH93782.1 | |
BC111943.1 | 12 | 1521 | AAI11944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss | RNF224 TPRN SLC34A3 NDOR1 RNF208 TMEM203 CYSRT1 |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss | TPRN NDOR1 RNF208 TMEM203 |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | ANAPC2 LRRC26 RNF224 TPRN RNF208 DPP7 LOC101930307 TMEM210 UAP1L1 MAN1B1 SSNA1 MIR3621 SLC34A3 NDOR1 MAN1B1-AS1 TMEM203 CYSRT1 GRIN1 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | ANAPC2 C9orf173-AS1 RNF224 TPRN STPG3 RNF208 NELFB EXD3 SSNA1 TOR4A NRARP SLC34A3 NDOR1 FAM166A TMEM203 CYSRT1 TUBB4B |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | ANAPC2 C9orf173-AS1 SAPCD2 TPRN RABL6 NOXA1 PTGDS LOC101930307 C9orf139 UAP1L1 MIR4292 SSNA1 PRR31 NRARP TMEM203 PHPT1 NSMF FBXW5 LRRC26 LCNL1 C9orf142 MAN1B1 EXD3 NPDC1 TRAF2 SLC34A3 NDOR1 ENTPD2 CLIC3 FUT7 CCDC183 RNF224 MIR4479 EDF1 RNF208 TMEM210 MIR3621 MAMDC4 ENTPD8 PNPLA7 GRIN1 TUBB4B C8G ABCA2 STPG3 DPP7 NELFB C9orf172 LCN12 TOR4A FAM166A MIR7114 MAN1B1-AS1 CCDC183-AS1 CYSRT1 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | ANAPC2 C9orf173-AS1 LRRC26 RNF224 TPRN STPG3 RNF208 TMEM210 NELFB EXD3 SSNA1 MIR3621 TOR4A NRARP SLC34A3 NDOR1 FAM166A TMEM203 CYSRT1 GRIN1 TUBB4B |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | ANAPC2 C9orf173-AS1 LRRC26 RNF224 TPRN STPG3 RNF208 NOXA1 TMEM210 NELFB EXD3 SSNA1 MIR3621 TOR4A NRARP SLC34A3 NDOR1 FAM166A ENTPD8 TMEM203 CYSRT1 GRIN1 TUBB4B |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | ANAPC2 C9orf173-AS1 MIR126 SAPCD2 TPRN LCN6 LINC01451 RABL6 PTGDS LOC101930307 C9orf139 UAP1L1 MIR4292 SSNA1 PRR31 FAM69B NRARP TMEM203 PHPT1 FBXW5 LRRC26 LCNL1 EGFL7 C9orf142 LCN8 MAN1B1 EXD3 NPDC1 TRAF2 SLC34A3 NDOR1 ENTPD2 CLIC3 FUT7 CCDC183 RNF224 SNHG7 MIR4479 EDF1 TMEM141 RNF208 TMEM210 SNORA17B MIR6722 MIR3621 MAMDC4 GRIN1 TUBB4B C8G ABCA2 STPG3 LCN10 DPP7 LOC100128593 SNORA17A AGPAT2 NELFB C9orf172 HSPC324 LCN12 TOR4A FAM166A LCN15 MAN1B1-AS1 CCDC183-AS1 CYSRT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |