Assay Details
Target Gene Details
Entrez Gene ID: | 55152 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:49015399-49021386 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DALRD3 | NM_001009996.2 | NP_001009996.1 | ||
NM_001276405.1 | NP_001263334.1 | |||
NM_018114.5 | NP_060584.3 | |||
XM_005265269.3 | 11 | 1594 | XP_005265326.1 | |
XM_006713219.2 | 12 | 1749 | XP_006713282.1 | |
XM_011533891.1 | XP_011532193.1 | |||
XM_011533892.1 | 11 | 1945 | XP_011532194.1 | |
XM_011533893.1 | 8 | 1703 | XP_011532195.1 | |
XM_017006723.1 | XP_016862212.1 | |||
AA553582.1 | 1 | 172 | ||
AK001358.1 | BAA91647.1 | |||
AK093054.1 | 2 | 2731 | ||
AK093204.1 | BAC04095.1 | |||
AK093294.1 | BAC04123.1 | |||
AK308273.1 | ||||
BC032440.1 | AAH32440.2 | |||
BC047683.1 | AAH47683.1 | |||
BC054493.1 | AAH54493.1 | |||
BX648719.1 | 6 | 1369 | ||
CF455269.1 |
Target Gene Details
Entrez Gene ID: | 11180 |
Gene Name: | WD repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.3:49007062-49015953 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WDR6 | NM_001320546.1 | 6 | 4409 | NP_001307475.1 |
NM_001320547.1 | 6 | 3780 | NP_001307476.1 | |
NM_018031.4 | 6 | 4195 | NP_060501.3 | |
AF099100.1 | 6 | 3864 | AAF80244.1 | |
AI636120.1 | 1 | 221 | ||
AK001080.1 | BAA91497.1 | |||
AK022719.1 | 5 | 2100 | BAB14201.1 | |
AK092079.1 | 6 | 1520 | ||
AK225653.1 | ||||
AL133589.1 | 5 | 1975 | CAB63730.1 | |
AL833910.1 | 5 | 3041 | CAD38766.1 | |
BC002826.2 | 8 | 1876 | AAH02826.1 | |
BC078176.1 | 5 | 1914 | AAH78176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion | P4HTM DALRD3 WDR6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |