Assay Details
Target Gene Details
Entrez Gene ID: | 23528 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
ZBP-99, ZNP-99 |
Location: |
Chr.1:200404947-200483604 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF281 | NM_001281293.1 | NP_001268222.1 | ||
NM_001281294.1 | NP_001268223.1 | |||
NM_012482.4 | NP_036614.1 | |||
XM_017000887.1 | 4 | 1422 | XP_016856376.1 | |
XM_017000888.1 | 4 | 380 | XP_016856377.1 | |
AJ132592.1 | CAB70968.1 | |||
AK027284.1 | ||||
AK094701.1 | ||||
BC060820.1 | AAH60820.1 | |||
BP216938.1 | ||||
DB150929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548990 | Chr.1:200400630 - 200452338 on Build GRCh38 | Loss | ZNF281 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |