Assay Details
Target Gene Details
Entrez Gene ID: | 23196 |
Gene Name: | family with sequence similarity 120A |
Gene Aliases: |
C9orf10, OSSA |
Location: |
Chr.9:93451696-93566115 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM120A | NM_001286722.1 | 1 | 566 | NP_001273651.1 |
NM_001286723.1 | 1 | 566 | NP_001273652.1 | |
NM_001286724.1 | 1 | 566 | NP_001273653.1 | |
NM_014612.4 | 1 | 566 | NP_055427.2 | |
XM_005251842.4 | 1 | 518 | XP_005251899.1 | |
XM_011518412.2 | 1 | 383 | XP_011516714.1 | |
XM_011518413.2 | 1 | 383 | XP_011516715.1 | |
XM_011518414.2 | 1 | 383 | XP_011516716.1 | |
XM_011518416.2 | 1 | 384 | XP_011516718.1 | |
AF214737.1 | 1 | 371 | AAF72866.1 | |
AF214738.1 | 1 | 371 | AAF72867.1 | |
BC098584.1 | 1 | 542 | AAH98584.1 | |
BC111736.1 | 1 | 364 | AAI11737.1 | |
D80005.2 | 1 | 327 | BAA11500.2 |
Target Gene Details
Entrez Gene ID: | 158293 |
Gene Name: | family with sequence similarity 120A opposite strand |
Gene Aliases: |
C9orf10OS |
Location: |
Chr.9:93446500-93453592 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM120AOS | NM_198841.2 | 1 | 1332 | NP_942138.2 |
NR_136229.1 | ||||
NR_136230.1 | ||||
NR_136231.1 | 1 | 1332 | ||
AK056096.1 | 1 | 1332 | ||
AK309269.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |