Assay Details
Target Gene Details
Entrez Gene ID: | 10499 |
Gene Name: | nuclear receptor coactivator 2 |
Gene Aliases: |
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75 |
Location: |
Chr.8:70109762-70405390 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCOA2 | NM_001321703.1 | 23 | 4759 | NP_001308632.1 |
NM_001321707.1 | 23 | 4605 | NP_001308636.1 | |
NM_001321711.1 | 22 | 4480 | NP_001308640.1 | |
NM_001321712.1 | 22 | 4397 | NP_001308641.1 | |
NM_001321713.1 | 22 | 4500 | NP_001308642.1 | |
NM_006540.3 | 23 | 4687 | NP_006531.1 | |
XM_017012961.1 | 23 | 4881 | XP_016868450.1 | |
XM_017012962.1 | 23 | 4905 | XP_016868451.1 | |
XM_017012963.1 | 23 | 6118 | XP_016868452.1 | |
XM_017012964.1 | 23 | 5173 | XP_016868453.1 | |
XM_017012965.1 | 23 | 4838 | XP_016868454.1 | |
XM_017012966.1 | 24 | 5102 | XP_016868455.1 | |
XM_017012967.1 | 22 | 4816 | XP_016868456.1 | |
XM_017012968.1 | 24 | 4903 | XP_016868457.1 | |
XM_017012969.1 | 22 | 4934 | XP_016868458.1 | |
XM_017012970.1 | 22 | 11353 | XP_016868459.1 | |
XM_017012971.1 | 22 | 5065 | XP_016868460.1 | |
XM_017012972.1 | 19 | 4396 | XP_016868461.1 | |
AK298528.1 | ||||
BC114383.1 | 22 | 4554 | ||
X97674.1 | 23 | 4632 | CAA66263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831352 | Chr.8:69999548 - 70182606 on Build GRCh38 | Gain | PRDM14 NCOA2 |
nsv6246 | Chr.8:70106516 - 70137048 on Build GRCh38 | Insertion | NCOA2 |
More Information
Additional Information:
For this assay, SNP(s) [rs76809412] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |