Assay Details
Target Gene Details
Entrez Gene ID: | 57162 |
Gene Name: | pellino E3 ubiquitin protein ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.2:64092652-64144471 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PELI1 | NM_020651.3 | 7 | 2846 | NP_065702.2 |
XM_005264437.3 | 6 | 2665 | XP_005264494.1 | |
XM_011532994.2 | 7 | 3505 | XP_011531296.1 | |
XM_011532996.2 | 7 | 4264 | XP_011531298.1 | |
XM_011532997.2 | 7 | 2881 | XP_011531299.1 | |
XM_017004520.1 | 8 | 2980 | XP_016860009.1 | |
XM_017004521.1 | 8 | 2839 | XP_016860010.1 | |
AF302505.1 | 6 | 6424 | AAG15393.1 | |
AK027668.1 | 4 | 2157 | BAB55280.1 | |
BC050019.1 | 7 | 2800 | AAH50019.1 | |
BC063611.1 | 7 | 2774 | AAH63611.1 | |
BX649152.1 | 3 | 3702 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv151e55 | Chr.2:64085008 - 64294232 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv582127 | Chr.2:63618350 - 64923736 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1013132 | Chr.2:64008041 - 64403950 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834243 | Chr.2:64089380 - 64249801 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
nsv428396 | Chr.2:64025871 - 64312422 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv582126 | Chr.2:63346276 - 64470570 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)