Assay Details
Target Gene Details
Entrez Gene ID: | 4848 |
Gene Name: | CCR4-NOT transcription complex subunit 2 |
Gene Aliases: |
CDC36, HSPC131, NOT2, NOT2H |
Location: |
Chr.12:70242993-70354993 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNOT2 | NM_001199302.1 | NP_001186231.1 | ||
NM_014515.5 | NP_055330.1 | |||
XM_006719429.3 | XP_006719492.1 | |||
XM_006719430.3 | XP_006719493.1 | |||
XM_006719432.3 | XP_006719495.1 | |||
XM_006719433.3 | XP_006719496.1 | |||
XM_011538400.2 | XP_011536702.1 | |||
XM_017019348.1 | XP_016874837.1 | |||
XM_017019349.1 | XP_016874838.1 | |||
XM_017019350.1 | XP_016874839.1 | |||
XM_017019351.1 | XP_016874840.1 | |||
XM_017019352.1 | XP_016874841.1 | |||
XM_017019353.1 | XP_016874842.1 | |||
XM_017019354.1 | XP_016874843.1 | |||
AK315715.1 | ||||
DA445519.1 | ||||
DB038192.1 |
Target Gene Details
Entrez Gene ID: | 101928062 |
Gene Name: | long intergenic non-protein coding RNA 1481 |
Gene Aliases: |
- |
Location: |
Chr.12:70222190-70243360 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01481 | NR_120459.1 | |||
NR_120460.1 | ||||
NR_120461.1 | ||||
CA418524.1 | ||||
DA349921.1 | ||||
DB462634.1 | ||||
HY002672.1 | ||||
HY013176.1 | ||||
HY048577.1 | ||||
HY064474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3629884 | Chr.12:70233284 - 70252849 on Build GRCh38 | Gain | CNOT2 LINC01481 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |