Assay Details
Target Gene Details
Entrez Gene ID: | 5156 |
Gene Name: | platelet derived growth factor receptor alpha |
Gene Aliases: |
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA |
Location: |
Chr.4:54229097-54298245 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFRA | NM_006206.4 | NP_006197.1 | ||
XM_005265743.1 | XP_005265800.1 | |||
XM_006714039.3 | XP_006714102.1 | |||
XM_006714041.3 | XP_006714104.1 | |||
XM_011534385.2 | XP_011532687.1 | |||
XM_017008280.1 | XP_016863769.1 | |||
XM_017008281.1 | XP_016863770.1 | |||
XM_017008282.1 | XP_016863771.1 | |||
AK308353.1 | ||||
AK311006.1 | ||||
AK316578.1 | ||||
BC063414.1 | AAH63414.1 | |||
M21574.1 | AAA96715.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |