Assay Details
Target Gene Details
Entrez Gene ID: | 55862 |
Gene Name: | ethylmalonyl-CoA decarboxylase 1 |
Gene Aliases: |
HEL-S-76, MMCD, dJ351K20.2 |
Location: |
Chr.6:127288712-127343609 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECHDC1 | NM_001002030.1 | 6 | 1070 | NP_001002030.1 |
NM_001105544.1 | 5 | 848 | NP_001099014.1 | |
NM_001105545.1 | 4 | 795 | NP_001099015.1 | |
NM_001139510.1 | 6 | 848 | NP_001132982.1 | |
NM_018479.3 | 5 | 1017 | NP_060949.2 | |
XM_005267047.2 | 7 | 995 | XP_005267104.1 | |
XM_005267048.1 | 6 | 1169 | XP_005267105.1 | |
XM_005267049.2 | 5 | 1284 | XP_005267106.1 | |
XM_005267050.2 | 6 | 910 | XP_005267107.1 | |
XM_011535942.2 | 7 | 966 | XP_011534244.1 | |
XM_017011056.1 | 7 | 986 | XP_016866545.1 | |
AF220192.1 | 5 | 1021 | AAF67657.1 | |
AK021464.1 | 2 | 1342 | ||
AK025760.1 | 6 | 820 | ||
AK025796.1 | 6 | 848 | ||
AK098146.1 | 1 | 1700 | ||
AK303812.1 | 5 | 616 | ||
AL834469.1 | 6 | 1009 | CAD39128.1 | |
BC003549.1 | 6 | 1032 | AAH03549.1 | |
BE896631.1 | 5 | 700 | ||
EU668355.1 | 4 | 795 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |