Assay Details
Target Gene Details
Entrez Gene ID: | 8564 |
Gene Name: | kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) |
Gene Aliases: |
dJ317G22.1 |
Location: |
Chr.1:241531883-241595647 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KMO | NM_003679.4 | 15 | 4904 | NP_003670.2 |
AF056032.1 | 15 | 4639 | AAC62615.1 | |
AI262990.1 | 1 | 354 | ||
DB373228.1 | 1 | 363 |
Target Gene Details
Entrez Gene ID: | 23596 |
Gene Name: | opsin 3 |
Gene Aliases: |
ECPN, PPP1R116 |
Location: |
Chr.1:241593150-241640399 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OPN3 | NM_014322.2 | NP_055137.2 | ||
AF140242.1 | AAD32671.1 | |||
AF303588.1 | AAK37447.1 | |||
AK310307.1 | ||||
BC036773.1 | AAH36773.1 | |||
BC062622.1 | AAH62622.1 | |||
BC073872.1 | AAH73872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain | FH WDR64 CHML KMO RGS7 OPN3 |
nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain | FH WDR64 EXO1 CHML KMO RGS7 OPN3 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |