Assay Details
Target Gene Details
Entrez Gene ID: | 50650 |
Gene Name: | Rho guanine nucleotide exchange factor 3 |
Gene Aliases: |
GEF3, STA3, XPLN |
Location: |
Chr.3:56727418-57079308 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF3 | NM_001128615.1 | 13 | 2986 | NP_001122087.1 |
NM_001128616.1 | 10 | 3402 | NP_001122088.1 | |
NM_001289698.1 | 11 | 2910 | NP_001276627.1 | |
NM_019555.2 | 10 | 2949 | NP_062455.1 | |
XM_005265186.4 | 13 | 3050 | XP_005265243.1 | |
XM_005265187.3 | 13 | 3330 | XP_005265244.1 | |
XM_011533764.1 | 12 | 2909 | XP_011532066.1 | |
XM_011533765.1 | 12 | 2897 | XP_011532067.1 | |
XM_011533766.2 | 12 | 3241 | XP_011532068.1 | |
XM_011533767.1 | 11 | 2861 | XP_011532069.1 | |
XM_017006502.1 | 15 | 3251 | XP_016861991.1 | |
XM_017006503.1 | 13 | 2937 | XP_016861992.1 | |
XM_017006504.1 | 13 | 2995 | XP_016861993.1 | |
XM_017006505.1 | 10 | 2892 | XP_016861994.1 | |
XM_017006506.1 | 10 | 3154 | XP_016861995.1 | |
AB209661.1 | 10 | 3342 | BAD92898.1 | |
AF249744.1 | 10 | 2907 | AAF79954.1 | |
AF433662.1 | 13 | 2943 | AAP97313.1 | |
AK024340.1 | 8 | 2571 | BAB14891.1 | |
AL136832.1 | 10 | 2884 | CAB66766.1 | |
AX775923.1 | 10 | 2884 | CAE11661.1 | |
BC054345.1 | 10 | 2897 | ||
BC068513.1 | 10 | 2931 | AAH68513.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |