Assay Details
Target Gene Details
Entrez Gene ID: | 9743 |
Gene Name: | Rho GTPase activating protein 32 |
Gene Aliases: |
GC-GAP, GRIT, PX-RICS, RICS, p200RhoGAP, p250GAP |
Location: |
Chr.11:128965060-129279168 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP32 | NM_001142685.1 | 22 | 6928 | NP_001136157.1 |
NM_014715.3 | 13 | 6260 | NP_055530.2 | |
XM_005271736.4 | 23 | 7958 | XP_005271793.2 | |
XM_011543072.2 | 23 | 7160 | XP_011541374.1 | |
XM_011543073.2 | 23 | 6896 | XP_011541375.2 | |
XM_011543075.2 | 14 | 6106 | XP_011541377.1 | |
XM_017018595.1 | 23 | 6877 | XP_016874084.1 | |
XM_017018596.1 | 21 | 7154 | XP_016874085.1 | |
AB018255.2 | 13 | 6181 | ||
AB088416.1 | 13 | 6181 | ||
AK023715.1 | 1 | 1726 | ||
AY194287.1 | 13 | 6184 | AAO43677.1 | |
EF127492.1 | 22 | 6928 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |