Assay Details
Target Gene Details
Entrez Gene ID: | 10231 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4 |
Location: |
Chr.6:46220730-46492067 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RCAN2 | NM_001251973.1 | 5 | 904 | NP_001238902.1 |
NM_001251974.1 | 5 | 857 | NP_001238903.1 | |
NM_005822.3 | 4 | 979 | NP_005813.2 | |
XM_011514226.1 | 5 | 946 | XP_011512528.1 | |
AK090990.1 | 5 | 828 | ||
AK312392.1 | ||||
AY034085.1 | 5 | 811 | AAK59805.1 | |
AY034086.1 | 5 | 824 | AAK59806.1 | |
BC038509.1 | 4 | 762 | AAH38509.1 | |
D83407.1 | 4 | 755 | BAA11911.1 | |
DA409014.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |