Assay Details
Target Gene Details
Entrez Gene ID: | 57680 |
Gene Name: | chromodomain helicase DNA binding protein 8 |
Gene Aliases: |
AUTS18, HELSNF1 |
Location: |
Chr.14:21385194-21437298 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHD8 | NM_001170629.1 | 33 | 6771 | NP_001164100.1 |
NM_020920.3 | 34 | 6016 | NP_065971.2 | |
AB046784.3 | 33 | 6259 | BAB13390.3 | |
AK056639.1 | 14 | 2638 | ||
AK131077.1 | 19 | 3595 | BAC85127.1 | |
BC011695.2 | 2 | 317 | AAH11695.2 | |
BC025964.1 | 2 | 317 | AAH25964.1 | |
BC036920.1 | AAH36920.1 | |||
BC063693.1 | 2 | 335 | AAH63693.1 | |
BC073903.1 | 3 | 441 | AAH73903.1 | |
BC098452.1 | 8 | 1687 | AAH98452.1 | |
CR749315.1 | 34 | 5961 | CAH18170.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |