Assay Details
Target Gene Details
Entrez Gene ID: | 84134 |
Gene Name: | translocase of outer mitochondrial membrane 40 like |
Gene Aliases: |
TOMM40B |
Location: |
Chr.1:161225939-161230746 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOMM40L | NM_001286373.1 | 9 | 1561 | NP_001273302.1 |
NM_001286374.1 | 8 | 1468 | NP_001273303.1 | |
NM_032174.5 | 10 | 1663 | NP_115550.2 | |
XM_006711572.2 | 10 | 1538 | XP_006711635.1 | |
XM_011510057.2 | 8 | 1231 | XP_011508359.1 | |
XM_017002481.1 | 10 | 1501 | XP_016857970.1 | |
XM_017002482.1 | 9 | 1429 | XP_016857971.1 | |
AK022832.1 | 7 | 1279 | BAB14265.1 | |
AK027704.1 | 10 | 1542 | BAB55309.1 | |
BC014946.1 | 9 | 1516 | AAH14946.1 | |
BX537951.1 | 10 | 1540 | CAD97918.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |