Assay Details
Target Gene Details
Entrez Gene ID: | 58504 |
Gene Name: | Rho GTPase activating protein 22 |
Gene Aliases: |
RhoGAP2, RhoGap22 |
Location: |
Chr.10:48429831-48656265 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950952 | Chr.10:48445358 - 48458757 on Build GRCh38 | Deletion |
![]() |
nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)