Assay Details
Target Gene Details
Entrez Gene ID: | 57211 |
Gene Name: | adhesion G protein-coupled receptor G6 |
Gene Aliases: |
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR |
Location: |
Chr.6:142301919-142446266 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRG6 | NM_001032394.2 | 1 | 130 | NP_001027566.1 |
NM_001032395.2 | 1 | 130 | NP_001027567.1 | |
NM_020455.5 | 1 | 130 | NP_065188.4 | |
NM_198569.2 | 1 | 130 | NP_940971.1 | |
XM_005267061.3 | XP_005267118.1 | |||
XM_006715516.2 | XP_006715579.1 | |||
XM_006715518.2 | XP_006715581.1 | |||
XM_017011085.1 | XP_016866574.1 | |||
AB183546.1 | 1 | 120 | BAD27571.1 | |
AB183547.1 | 1 | 120 | BAD27572.1 | |
AB183548.1 | 1 | 120 | BAD27573.1 | |
AB183549.1 | 1 | 120 | BAD27574.1 | |
AF216967.1 | 1 | 135 | AAO13250.1 | |
AK122617.1 | ||||
AK307543.1 | ||||
BC075798.1 | 1 | 190 | AAH75798.1 | |
BX640971.1 | CAE45986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830825 | Chr.6:142264105 - 142431438 on Build GRCh38 | Gain | ADGRG6 |
nsv1029666 | Chr.6:142272962 - 142321947 on Build GRCh38 | Gain | ADGRG6 |
esv275340 | Chr.6:142299268 - 142303431 on Build GRCh38 | Gain+Loss | ADGRG6 |
More Information
Additional Information:
For this assay, SNP(s) [rs73576330] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |