Assay Details
Target Gene Details
Entrez Gene ID: | 81491 |
Gene Name: | G protein-coupled receptor 63 |
Gene Aliases: |
PSP24(beta), PSP24B |
Location: |
Chr.6:96794122-96837477 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR63 | NM_001143957.2 | 3 | 1340 | NP_001137429.1 |
NM_030784.3 | 2 | 1222 | NP_110411.1 | |
XM_006715570.3 | 2 | 1292 | XP_006715633.1 | |
XM_011536153.2 | 3 | 1283 | XP_011534455.1 | |
XM_011536154.2 | 3 | 1156 | XP_011534456.1 | |
XM_011536155.2 | 3 | 1335 | XP_011534457.1 | |
XM_011536157.2 | 2 | 1058 | XP_011534459.1 | |
XM_011536159.2 | 2 | 1227 | XP_011534461.1 | |
XM_017011334.1 | 2 | 1750 | XP_016866823.1 | |
AB030566.1 | 2 | 1069 | BAB20031.1 | |
AB209347.1 | 2 | 1208 | BAD92584.1 | |
AK289839.1 | 2 | 1222 | ||
AK313535.1 | 2 | 1191 | ||
BC067465.1 | 1 | 897 | AAH67465.1 | |
BC067466.1 | 1 | 898 | AAH67466.1 | |
BC067467.1 | 1 | 898 | AAH67467.1 | |
BC067468.1 | 1 | 898 | AAH67468.1 | |
BC067469.1 | 1 | 898 | AAH67469.1 | |
FJ348260.1 | 1 | 862 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |