Assay Details
Target Gene Details
Entrez Gene ID: | 285429 |
Gene Name: | DDB1 and CUL4 associated factor 4 like 1 |
Gene Aliases: |
WDR21B |
Location: |
Chr.4:41981696-41986467 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF4L1 | NM_001029955.3 | 1 | 120 | NP_001025126.2 |
AK122914.1 | ||||
BC035027.1 | AAH35027.1 | |||
BC052943.1 | AAH52943.1 | |||
BC104224.1 | AAI04225.1 | |||
BC104225.1 | AAI04226.1 | |||
DB100231.1 | 1 | 120 |