Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | 2 | 305 | NP_073741.2 |
AB101208.1 | 2 | 219 | ||
AK026000.1 | 2 | 212 | BAB15314.1 | |
AK292610.1 | 2 | 304 | ||
BC110910.1 | 2 | 194 | AAI10911.1 | |
BC128263.1 | 2 | 198 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | EML3 ROM1 MTA2 TUT1 B3GAT3 MIR6747 GANAB AHNAK EEF1G |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | MTA2 TUT1 MIR6747 AHNAK EEF1G |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | EML3 ROM1 MTA2 TUT1 B3GAT3 MIR6747 GANAB EEF1G |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | UBXN1 MTA2 SNORA57 INTS5 BSCL2 METTL12 LBHD1 EEF1G LRRN4CL EML3 ROM1 TUT1 B3GAT3 MIR6747 GANAB C11orf98 AHNAK HNRNPUL2-BSCL2 UQCC3 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | UBXN1 MTA2 SNORA57 INTS5 BSCL2 METTL12 LBHD1 EEF1G LRRN4CL EML3 ROM1 GNG3 TUT1 B3GAT3 MIR6747 GANAB HNRNPUL2 TTC9C C11orf98 HNRNPUL2-BSCL2 UQCC3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |