Assay Details
Target Gene Details
Entrez Gene ID: | 23635 |
Gene Name: | single stranded DNA binding protein 2 |
Gene Aliases: |
HSPC116, SOSS-B2 |
Location: |
Chr.5:81417360-81751253 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SSBP2 | NM_001256732.1 | 17 | 1517 | NP_001243661.1 |
NM_001256733.1 | 16 | 1433 | NP_001243662.1 | |
NM_001256734.1 | 16 | 1427 | NP_001243663.1 | |
NM_001256735.1 | 16 | 1403 | NP_001243664.1 | |
NM_001256736.1 | 15 | 1304 | NP_001243665.1 | |
NM_012446.3 | 17 | 1493 | NP_036578.2 | |
XM_011543297.2 | 16 | 1333 | XP_011541599.1 | |
XM_017009304.1 | 17 | 1335 | XP_016864793.1 | |
XM_017009305.1 | 18 | 1424 | XP_016864794.1 | |
XM_017009306.1 | 16 | 1309 | XP_016864795.1 | |
XM_017009307.1 | 16 | 1221 | XP_016864796.1 | |
XM_017009308.1 | 18 | 1596 | XP_016864797.1 | |
XM_017009309.1 | 17 | 1660 | XP_016864798.1 | |
XM_017009310.1 | 17 | 1808 | XP_016864799.1 | |
AF077048.1 | AAD27781.1 | |||
AF161465.1 | 17 | 1323 | AAF29080.1 | |
AK074537.1 | 17 | 1288 | BAC11046.1 | |
AK293544.1 | 16 | 1228 | ||
AK294864.1 | 15 | 1099 | ||
AK297494.1 | 17 | 1201 | ||
AK299858.1 | 16 | 1271 | ||
AL080076.1 | 17 | 1341 | CAB45699.1 | |
BC017020.1 | 17 | 1341 | AAH17020.1 | |
BX397789.2 | 1 | 406 | ||
BX413405.2 | 1 | 393 | ||
CR936787.1 | 16 | 1248 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3605594 | Chr.5:81406976 - 81473957 on Build GRCh38 | Gain | SSBP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |