Assay Details
Target Gene Details
Entrez Gene ID: | 2334 |
Gene Name: | AF4/FMR2 family member 2 |
Gene Aliases: |
FMR2, FMR2P, FRAXE, MRX2, OX19 |
Location: |
Chr.X:148500619-149000663 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AFF2 | NM_001169122.1 | NP_001162593.1 | ||
NM_001169123.1 | NP_001162594.1 | |||
NM_001169124.1 | NP_001162595.1 | |||
NM_001169125.1 | NP_001162596.1 | |||
NM_002025.3 | NP_002016.2 | |||
AB102644.1 | BAC81113.1 | |||
AJ001550.1 | CAA04822.1 | |||
BC132683.1 | ||||
BC143740.1 | ||||
BC143744.1 | ||||
L76569.1 | AAA99416.1 | |||
U48436.1 | AAC82513.1 | |||
X95463.1 | CAA64730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523386 | Chr.X:148473389 - 148517170 on Build GRCh38 | Gain | AFF2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |