Assay Details
Target Gene Details
Entrez Gene ID: | 26523 |
Gene Name: | argonaute 1, RISC catalytic component |
Gene Aliases: |
EIF2C, EIF2C1, GERP95, Q99, hAgo1 |
Location: |
Chr.1:35869808-35929610 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGO1 | NM_001317122.1 | 4 | 685 | NP_001304051.1 |
NM_001317123.1 | 4 | 529 | NP_001304052.1 | |
NM_012199.4 | 4 | 685 | NP_036331.1 | |
XM_011541236.2 | 4 | 646 | XP_011539538.1 | |
XM_017001016.1 | 4 | 646 | XP_016856505.1 | |
XM_017001017.1 | 4 | 646 | XP_016856506.1 | |
XM_017001018.1 | XP_016856507.1 | |||
XM_017001019.1 | 4 | 661 | XP_016856508.1 | |
AF093097.1 | 4 | 671 | AAF00068.1 | |
AK001622.1 | 4 | 529 | ||
AK314148.1 | 4 | 646 | ||
BC063275.1 | 4 | 665 | AAH63275.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |