Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD10 | NM_001317395.1 | 7 | 2772 | NP_001304324.1 |
NM_001317399.1 | 7 | 2700 | NP_001304328.1 | |
NM_031954.4 | 7 | 2769 | NP_114160.1 | |
NR_133897.1 | 6 | 2599 | ||
NR_133898.1 | 6 | 2555 | ||
AB209358.1 | 2 | 5314 | BAD92595.1 | |
AF113208.1 | 7 | 2514 | AAG39279.1 | |
AK027543.1 | 7 | 2575 | BAB55188.1 | |
AK092170.1 | 6 | 2388 | ||
AK123733.1 | 4 | 3006 | ||
AY205299.1 | 7 | 2575 | AAO47716.1 | |
BC040062.1 | 7 | 2522 | AAH40062.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |